5-Hydroxypyrimidin-4(1H)-one ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Hydroxypyrimidin-4(3H)-one
A212633-1G
A212633-5G
77883-512EA
121
USD
77883-512
77883-513
5-Hydroxypyrimidin-4(1H)-one ≥98%
5-Hydroxypyrimidin-4(1H)-one
Precaución: for Research Use Only
Formula:
C4H4N2O2 |
MDL Number:
MFCD11656602 CAS Number: 15837-41-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...