(S)-4-Benzyl-3-(3-methylbutanoyl)oxazolidin-2-one ≥97%

Supplier: Ambeed
Warning

A212537-1G A212537-5G
77885-521EA 18 USD
77885-521 77885-522
(S)-4-Benzyl-3-(3-methylbutanoyl)oxazolidin-2-one ≥97%
(S)-4-Benzyl-3-(3-methylbutanoyl)oxazolidin-2-one

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₃
MDL Number: MFCD09817644
CAS Number: 104266-90-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR