Cefditoren pivoxil ≥98%

Supplier: Ambeed
Warning

Synonyms: (6R,7R)-(pivaloyloxy)methyl 7-((Z)-2-(2-aminothiazol-4-yl)-2-(methoxyimino)acetamido)-3-((Z)-2-(4-methylthiazol-5-yl)vinyl)-8-oxo-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylate

A212367-10MG A212367-1MG A212367-5MG
77600-704EA 8.2 USD
77600-704 77600-706 77600-708
Cefditoren pivoxil ≥98%
Cefditoren pivoxil

Precaución: for Research Use Only

Formula: C₂₅H₂₈N₆O₇S₃
MW: 620.71 g/mol
MDL Number: MFCD00933166
CAS Number: 117467-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR