Sabutoclax ≥98%

Supplier: Ambeed
Warning

Synonyms: (1R)-1,1',6,6',7,7'-Hexahydroxy-3,3'-dimethyl-N5,N5'-bis((2R)-2-phenylpropyl)-[2,2'-binaphthalene]-5,5'-dicarboxamide

A211104-10MG A211104-1MG A211104-5MG
77615-788EA 575.77 USD
77615-788 77615-790 77615-792
Sabutoclax ≥98%
Sabutoclax

Precaución: for Research Use Only

Formula: C₄₂H₄₀N₂O₈
MW: 700.78 g/mol
MDL Number: MFCD26142645
CAS Number: 1228108-65-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR