Pyrrolidin-2-ylacetic acid hydrochloride ≥97%

Supplier: Ambeed
Warning

Synonyms: Pyrrolidin-2-yl-acetic acid hydrochloride

A211014-250MG A211014-1G
77612-078EA 26.78 USD
77612-078 77671-882
Pyrrolidin-2-ylacetic acid hydrochloride ≥97%
Pyrrolidin-2-ylacetic acid hydrochloride

Precaución: for Research Use Only

Formula: C6H12ClNO2
MW: 165.62 g/mol
MDL Number: MFCD11506301
CAS Number: 71985-79-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR