4,5-Dibromo-2-thenoic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: 4,5-Dibromo-2-thiophenecarboxylic acid, 4,5-Dibromothiophene-2-carboxylic acid

A210869-1G A210869-250MG
77612-074EA 6.18 USD
77612-074 77683-600
4,5-Dibromo-2-thenoic acid ≥98%
4,5-Dibromo-2-thenoic acid

Precaución: For research use only.

Formula: C₅H₂Br₂O₂S
MW: 285.94 g/mol
MDL Number: MFCD00039682
CAS Number: 6324-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR