5-Amino-1-(tert-butyl)-1H-pyrazol-3-ol ≥95%

Supplier: Ambeed
Warning

A210752-1G A210752-250MG A210752-5G
77671-876EA 32 USD
77671-876 77671-878 77671-880
5-Amino-1-(tert-butyl)-1H-pyrazol-3-ol ≥95%
5-Amino-1-(tert-butyl)-1H-pyrazol-3-ol

Precaución: for Research Use Only

Formula: C₇H₁₃N₃O
MDL Number: MFCD14607822
CAS Number: 359867-35-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR