VX-661 ≥99%

Supplier: Ambeed
Warning

Synonyms: Tezacaftor, (R)-1-(2,2-difluorobenzo[d][1,3]dioxol-5-yl)-N-(1-(2,3-dihydroxypropyl)-6-fluoro-2-(1-hydroxy-2-methylpropan-2-yl)-1H-indol-5-yl)cyclopropanecarboxamide

A210449-10MG A210449-1MG A210449-5MG A210449-250MG
77612-062EA 23.69 USD
77612-062 77612-064 77612-066 77685-549
VX-661 ≥99%
VX-661

Precaución: for Research Use Only

Formula: C₂₆H₂₇F₃N₂O₆
MW: 520.50 g/mol
MDL Number: MFCD23106064
CAS Number: 1152311-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR