5-(4-(Methylsulfonyl)phenyl)-6'-(trifluoromethyl)-[3,3'-bipyridin]-2-amine ≥98%

Supplier: Ambeed
Warning

Synonyms: MMV390048

A210038-10MG A210038-1MG A210038-5MG
77615-768EA 62.83 USD
77615-768 77615-770 77615-772
5-(4-(Methylsulfonyl)phenyl)-6'-(trifluoromethyl)-[3,3'-bipyridin]-2-amine ≥98%
5-(4-(Methylsulfonyl)phenyl)-6'-(trifluoromethyl)-[3,3'-bipyridin]-2-amine

Precaución: for Research Use Only

Formula: C₁₈H₁₄F₃N₃O₂S
MW: 393.38 g/mol
MDL Number: MFCD28502126
CAS Number: 1314883-11-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR