2-(3-Formylphenyl)acetic acid ≥97%

Supplier: Ambeed
Warning

A209963-100MG A209963-1G A209963-250MG
77890-251EA 42 USD
77890-251 77890-252 77890-253
2-(3-Formylphenyl)acetic acid ≥97%
2-(3-Formylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₈O₃
MDL Number: MFCD20638515
CAS Number: 34956-29-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR