3'-Methyl-2,2,2-trifluoroacetophenone ≥98%

Supplier: Ambeed
Warning

A209515-25G A209515-5G
77883-503EA 271 USD
77883-503 77883-504
3'-Methyl-2,2,2-trifluoroacetophenone ≥98%
3'-Methyl-2,2,2-trifluoroacetophenone

Precaución: for Research Use Only

Formula: C₉H₇F₃O
MDL Number: MFCD00461893
CAS Number: 1736-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR