tert-Butyl (4-bromo-3,5-dimethylphenyl)carbamate ≥98%

Supplier: Ambeed
Warning

A209410-1G A209410-250MG
77885-510EA 50 USD
77885-510 77885-511
tert-Butyl (4-bromo-3,5-dimethylphenyl)carbamate ≥98%
tert-Butyl (4-bromo-3,5-dimethylphenyl)carbamate

Precaución: for Research Use Only

Formula: C₁₃H₁₈BrNO₂
MDL Number: MFCD17214234
CAS Number: 361436-27-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR