1-Boc-(3-carboxymethoxy)azetidine ≥98%

Supplier: Ambeed
Warning

Synonyms: 1-Boc-(3-Carboxymethoxy)azetidine

A209170-100MG A209170-250MG A209170-1G A209170-5G
77612-050EA 47.38 USD
77612-050 77612-052 77671-854 77671-856
1-Boc-(3-carboxymethoxy)azetidine ≥98%
1-Boc-(3-carboxymethoxy)azetidine

Precaución: for Research Use Only

Formula: C₁₀H₁₇NO₅
MW: 231.250 g/mol
MDL Number: MFCD02179034
CAS Number: 889952-83-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR