(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride ≥98%

Supplier: Ambeed
Warning

A208602-1G A208602-100MG A208602-250MG
77671-850EA 173 USD
77671-850 77881-497 77881-498
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride ≥98%
(1R)-1-(4-Chloro-3-fluorophenyl)ethylamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₉ClFN·ClH
MDL Number: MFCD12547495
CAS Number: 1253790-80-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR