(5S,8S)-1-(9H-Fluoren-9-yl)-5-isopropyl-8-methyl-3,6,9-trioxo-2-oxa-4,7,10-triazaundecan-11-yl acetate ≥95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Val-Ala-aminomethyl acetate

A2085939-10MG A2085939-5MG A2085939-100MG A2085939-25MG A2085939-50MG
77626-056EA 182.31 USD
77626-056 77626-058 77877-651 77877-652 77877-653
(5S,8S)-1-(9H-Fluoren-9-yl)-5-isopropyl-8-methyl-3,6,9-trioxo-2-oxa-4,7,10-triazaundecan-11-yl acetate ≥95%
(5S,8S)-1-(9H-Fluoren-9-yl)-5-isopropyl-8-methyl-3,6,9-trioxo-2-oxa-4,7,10-triazaundecan-11-yl acetate

Precaución: for Research Use Only

Formula: C₂₆H₃₁N₃O₆
MW: 481.54 g/mol
CAS Number: 2505045-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR