(S)-2-Amino-2-(thiophen-2-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: H-D-α-(2-Thienyl)-Gly-OH

A208593-1G A208593-5G A208593-250MG
77612-048EA 306.94 USD
77612-048 77671-848 77671-846
(S)-2-Amino-2-(thiophen-2-yl)acetic acid ≥98%
(S)-2-Amino-2-(thiophen-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₂S
MW: 157.19 g/mol
MDL Number: MFCD00078081
CAS Number: 43189-45-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR