(R)-2-Methyl-N-(2-methyl-4-(N-(1-(1-methylpiperidin-4-yl)ethyl)sulfamoyl)phenyl)benzamide ≥95%

Supplier: Ambeed
Warning

A2085371-10MG A2085371-1MG A2085371-5MG
77598-304EA 215.25 USD
77598-304 77598-306 77598-308
(R)-2-Methyl-N-(2-methyl-4-(N-(1-(1-methylpiperidin-4-yl)ethyl)sulfamoyl)phenyl)benzamide ≥95%
(R)-2-Methyl-N-(2-methyl-4-(N-(1-(1-methylpiperidin-4-yl)ethyl)sulfamoyl)phenyl)benzamide

Precaución: for Research Use Only

Formula: C₂₃H₃₁N₃O₃S
CAS Number: 2756350-91-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR