Tetramethylammonium (trifluoromethyl)sulfanide ≥98%

Supplier: Ambeed
Danger

Synonyms: Tetramethylammonium trifluoromethanethiolate, Tetramethylazanium trifluoromethanethiolate

A207961-1G A207961-5G
77748-304EA 335 USD
77748-304 77748-305
Tetramethylammonium (trifluoromethyl)sulfanide ≥98%
Tetramethylammonium (trifluoromethyl)sulfanide

Precaución: for Research Use Only

Formula: CF₃S·C₄H₁₂N
MDL Number: MFCD24842708
CAS Number: 515823-30-0
UN: 1759
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR