(5-Bromopyridin-3-yl)-morpholin-4-yl-methanone ≥98%

Supplier: Ambeed
Warning

A207864-1G A207864-5G
77883-495EA 28 USD
77883-495 77883-496
(5-Bromopyridin-3-yl)-morpholin-4-yl-methanone ≥98%
(5-Bromopyridin-3-yl)-morpholin-4-yl-methanone

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrN₂O₂
MDL Number: MFCD01788376
CAS Number: 342013-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR