(-)-Quinine hydrochloride ≥99%

Supplier: Ambeed
Warning

Synonyms: (R)-(6-Methoxyquinolin-4-yl)((1S,2S,4S,5R)-5-vinylquinuclidin-2-yl)methanol hydrochloride, Quinine monohydrochloride

A207825-25G A207825-100G
77600-684EA 45.1 USD
77600-684 77667-746
(-)-Quinine hydrochloride ≥99%
(-)-Quinine hydrochloride

Precaución: for Research Use Only

Formula: C₂₀H₂₅ClN₂O₂
MW: 360.88 g/mol
MDL Number: MFCD00078498
CAS Number: 130-89-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR