5-Bromo-1,2,3,4-tetrahydronaphthalene ≥98%
Supplier: Ambeed
|
Warning
|
A207450-1G
A207450-250MG
77671-834EA
220
USD
77671-834
77671-836
5-Bromo-1,2,3,4-tetrahydronaphthalene ≥98%
5-Bromo-1,2,3,4-tetrahydronaphthalene
Precaución: for Research Use Only
Formula:
C₁₀H₁₁Br |
MDL Number:
MFCD08361743 CAS Number: 6134-55-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...