2,3,5,6,7,8-Hexahydro-1H-cyclopenta[b]quinolin-9-amine ≥97%

Supplier: Ambeed
Warning

Synonyms: Ipidacrine

A207110-1G A207110-250MG
77612-034EA 44.29 USD
77612-034 77612-036
2,3,5,6,7,8-Hexahydro-1H-cyclopenta[b]quinolin-9-amine ≥97%
2,3,5,6,7,8-Hexahydro-1H-cyclopenta[b]quinolin-9-amine

Precaución: for Research Use Only

Formula: C₁₂H₁₆N₂
MW: 188.26 g/mol
MDL Number: MFCD00618424
CAS Number: 62732-44-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR