6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one ≥98%

Supplier: Ambeed
Warning

Synonyms: 6-Bromo-3-methyl-1,3-benzoxazol-2(3H)-one, 6-Bromo-3-methylbenzo[d]oxazol-2(3H)-one

A206950-100MG A206950-250MG
77683-583EA 13 USD
77683-583 77683-584
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one ≥98%
6-Bromo-3-methyl-2,3-dihydro-1,3-benzoxazol-2-one

Precaución: For research use only.

Formula: C₈H₆BrNO₂
MW: 228.05 g/mol
MDL Number: MFCD00694789
CAS Number: 67927-44-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR