Bufalin ≥98%

Supplier: Ambeed
Danger

A206803-5MG A206803-10MG A206803-1MG
77615-730EA 52.31 USD
77615-730 77748-299 77748-300
Bufalin ≥98%
Bufalin

Precaución: for Research Use Only

Formula: C₂₄H₃₄O₄
MDL Number: MFCD00056525
CAS Number: 465-21-4
UN: 3462
ADR: 6.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR