(S)-Chroman-4-amine ≥97%

Supplier: Ambeed
Warning

A206694-100MG A206694-250MG
77872-041EA 33 USD
77872-041 77872-042
(S)-Chroman-4-amine ≥97%
(S)-Chroman-4-amine

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD06761755
CAS Number: 188198-38-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR