6-Bromo-2-phenyl-1H-indole ≥96%

Supplier: Ambeed
Warning

A206648-1G A206648-100MG A206648-5G
77612-024EA 198.79 USD
77612-024 77885-498 77885-499
6-Bromo-2-phenyl-1H-indole ≥96%
6-Bromo-2-phenyl-1H-indole

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrN
MDL Number: MFCD11976327
CAS Number: 77185-71-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR