N1,N3-Bis(2,3-dihydroxypropyl)-5-(2-hydroxy-N-(2-hydroxyethyl)acetamido)-2,4,6-triiodoisophthalamide ≥98%

Supplier: Ambeed
Warning

Synonyms: Ioversol

A206124-1G A206124-250MG A206124-5G
77879-284EA 21 USD
77879-284 77879-285 77879-286
N1,N3-Bis(2,3-dihydroxypropyl)-5-(2-hydroxy-N-(2-hydroxyethyl)acetamido)-2,4,6-triiodoisophthalamide ≥98%
N1,N3-Bis(2,3-dihydroxypropyl)-5-(2-hydroxy-N-(2-hydroxyethyl)acetamido)-2,4,6-triiodoisophthalamide

Precaución: for Research Use Only

Formula: C₁₈H₂₄I₃N₃O₉
MW: 807.11 g/mol
MDL Number: MFCD00867959
CAS Number: 87771-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR