Altiratinib ≥98%

Supplier: Ambeed
Warning

Synonyms: N-[4-[[2-[(Cyclopropylcarbonyl)amino]-4-pyridinyl]oxy]-2,5-difluorophenyl]-N′-(4-fluorophenyl)-1,1-cyclopropanedicarboxamide

A206114-100MG A206114-10MG A206114-5MG
77685-528EA 315 USD
77685-528 77685-529 77685-530
Altiratinib ≥98%
Altiratinib

Precaución: for Research Use Only

Formula: C₂₆H₂₁F₃N₄O₄
MW: 510.46 g/mol
MDL Number: MFCD28900672
CAS Number: 1345847-93-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR