tert-Butyl-2-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

A206027-1G A206027-250MG A206027-5G
77667-724EA 43 USD
77667-724 77667-726 77667-728
tert-Butyl-2-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate ≥95%
tert-Butyl-2-bromo-6,7-dihydrothieno[3,2-c]pyridine-5(4H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₆BrNO₂S
MW: 318.2300 g/mol
MDL Number: MFCD09878703
CAS Number: 949922-62-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR