2-Azaspiro[3.5]nonane hydrochloride ≥98%

Supplier: Ambeed
Warning

A205946-100MG A205946-1G
77892-295EA 79 USD
77892-295 77892-296
2-Azaspiro[3.5]nonane hydrochloride ≥98%
2-Azaspiro[3.5]nonane hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₆ClN
MDL Number: MFCD18914329
CAS Number: 1303968-07-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR