tert-Butyl 6-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A205868-250MG A205868-1G A205868-5G A205868-100MG
77612-018EA 53.56 USD
77612-018 77683-580 77683-581 77890-242
tert-Butyl 6-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate ≥98%, Ambeed.Inc
tert-Butyl 6-oxo-2-azabicyclo[2.2.1]heptane-2-carboxylate

Precaución: For research use only.

Formula: C₁₁H₁₇NO₃
MDL Number: MFCD13806381
CAS Number: 198835-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR