1,8-Diazaspiro[4.5]decane dihydrochloride ≥97%

Supplier: Ambeed
Warning

A205613-100MG A205613-1G A205613-250MG
77890-239EA 57 USD
77890-239 77890-240 77890-241
1,8-Diazaspiro[4.5]decane dihydrochloride ≥97%
1,8-Diazaspiro[4.5]decane dihydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₈Cl₂N₂
MDL Number: MFCD11501293
CAS Number: 1159822-20-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR