2-Amino-6,7-dihydrobenzo[d]thiazol-4(5H)-one ≥97%

Supplier: Ambeed
Warning

A205611-1G A205611-5G
77615-728EA 95.79 USD
77615-728 77746-269
2-Amino-6,7-dihydrobenzo[d]thiazol-4(5H)-one ≥97%
2-Amino-6,7-dihydrobenzo[d]thiazol-4(5H)-one

Precaución: for Research Use Only

Formula: C₇H₈N₂OS
MW: 168.22 g/mol
MDL Number: MFCD21117211
CAS Number: 36234-66-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR