3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid hydrochloride ≥95%

Supplier: Ambeed
Warning

A205562-100MG A205562-1G A205562-250MG A205562-5G
77612-006EA 28.16 USD
77612-006 77612-008 77612-010 77612-012
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid hydrochloride ≥95%
3-Aminobicyclo[1.1.1]pentane-1-carboxylic acid hydrochloride

Precaución: for Research Use Only

Formula: C₆H₉NO₂·ClH
MDL Number: MFCD09971722
CAS Number: 1172097-47-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR