2-(Trifluoromethyl)-5-pyridineboronic acid pinacol ester ≥98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2-Trifluoromethylpyridine-5-boronic acid pinacol ester, 2-(Trifluoromethyl)pyridine-5-boronic Acid Pinacol Ester
A205389-1G
A205389-250MG
A205389-25G
A205389-5G
77671-816EA
98
USD
77671-816
77883-485
77883-486
77883-487
2-(Trifluoromethyl)-5-pyridineboronic acid pinacol ester ≥98%
2-(Trifluoromethyl)-5-pyridineboronic acid pinacol ester
Precaución: for Research Use Only
Formula:
C₁₂H₁₅BF₃NO₂ MW: 273.592 g/mol |
MDL Number:
MFCD11226774 CAS Number: 1218790-39-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...