3,5-Diethynylpyridine ≥95%

Supplier: Ambeed
Warning

A205190-1G A205190-250MG
77883-483EA 118 USD
77883-483 77883-484
3,5-Diethynylpyridine ≥95%
3,5-Diethynylpyridine

Precaución: for Research Use Only

Formula: C₉H₅N
MDL Number: MFCD11109669
CAS Number: 67227-90-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR