1-(2-Bromophenyl)-3-(4-cyano-1H-benzo[d][1,2,3]triazol-7-yl)urea ≥99%

Supplier: Ambeed
Warning

A205049-1MG A205049-5MG A205049-10MG
77615-722EA 61.8 USD
77615-722 77615-724 77615-720
1-(2-Bromophenyl)-3-(4-cyano-1H-benzo[d][1,2,3]triazol-7-yl)urea ≥99%
1-(2-Bromophenyl)-3-(4-cyano-1H-benzo[d][1,2,3]triazol-7-yl)urea

Precaución: for Research Use Only

Formula: C₁₄H₉BrN₆O
MDL Number: MFCD09971124
CAS Number: 211096-49-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR