5-(2-Ethoxy-5-((4-(2-hydroxyethyl)piperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one ≥95%

Supplier: Ambeed
Warning

Synonyms: Lodenafil

A204453-10MG A204453-1MG A204453-5MG
77611-992EA 43.26 USD
77611-992 77611-994 77611-996
5-(2-Ethoxy-5-((4-(2-hydroxyethyl)piperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one ≥95%
5-(2-Ethoxy-5-((4-(2-hydroxyethyl)piperazin-1-yl)sulfonyl)phenyl)-1-methyl-3-propyl-1H-pyrazolo[4,3-d]pyrimidin-7(6H)-one

Precaución: for Research Use Only

Formula: C₂₃H₃₂N₆O₅S
MW: 504.60 g/mol
MDL Number: MFCD00908400
CAS Number: 139755-85-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR