2-(4-(Dimethylamino)styryl)-1-methylpyridin-1-ium iodide ≥97%

Supplier: Ambeed
Warning

A203657-5G A203657-1G A203657-250MG
77748-287EA 398 USD
77748-287 77748-285 77748-286
2-(4-(Dimethylamino)styryl)-1-methylpyridin-1-ium iodide ≥97%
2-(4-(Dimethylamino)styryl)-1-methylpyridin-1-ium iodide

Precaución: for Research Use Only

Formula: C₁₆H₁₉IN₂
MDL Number: MFCD00011980
CAS Number: 2156-29-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR