4-Hydroxypyridazine ≥97%

Supplier: Ambeed
Warning

A203175-10G A203175-1G A203175-5G
77748-282EA 428 USD
77748-282 77748-283 77748-284
4-Hydroxypyridazine ≥97%
4-Hydroxypyridazine

Precaución: for Research Use Only

Formula: C₄H₄N₂O
MW: 96.874 g/mol
MDL Number: MFCD00233967
CAS Number: 20733-10-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR