(3R,5aS,6R,8aS,9R,12R,12aR)-Decahydro-3,6,9-trimethyl-3,12-Epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10-ol mixture of alpha- and beta-isomers 95% mix

Supplier: Ambeed
Warning

A202607-100G A202607-5G A202607-25G A202607-10G A202607-1G
77593-316EA 442.8 USD
77593-316 77593-318 77615-706 77748-277 77748-278
(3R,5aS,6R,8aS,9R,12R,12aR)-Decahydro-3,6,9-trimethyl-3,12-Epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10-ol mixture of alpha- and beta-isomers 95% mix
(3R,5aS,6R,8aS,9R,12R,12aR)-Decahydro-3,6,9-trimethyl-3,12-Epoxy-12H-pyrano[4,3-j]-1,2-benzodioxepin-10-ol

Precaución: for Research Use Only

Formula: C₁₅H₂₄O₅
MW: 284.35 g/mol
MDL Number: MFCD28384156
CAS Number: 131175-87-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR