A83-01 ≥95%

Supplier: Ambeed
Warning

A202409-10MG A202409-1MG A202409-5MG
77611-954EA 79.46 USD
77611-954 77611-956 77611-958
A83-01 ≥95%
A83-01

Precaución: for Research Use Only

Formula: C₂₅H₁₉N₅S
MDL Number: MFCD08705403
CAS Number: 909910-43-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR