4-Pyridin-4-ylbutyric acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 4-(Pyridin-4-yl)butanoic acid

A201904-100MG A201904-1G
77746-258EA 26 USD
77746-258 77746-259
4-Pyridin-4-ylbutyric acid ≥97%
4-Pyridin-4-ylbutyric acid

Precaución: for Research Use Only

Formula: C₉H₁₁NO₂
MW: 165.19 g/mol
MDL Number: MFCD00673154
CAS Number: 102878-73-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR