7-Chloro-DL-tryptophan ≥95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(7-chloro-1H-indol-3-yl)propanoic acid, H-DL-Trp(7-Cl)-OH

A201790-1G A201790-100MG A201790-250MG A201790-5G
77683-563EA 248 USD
77683-563 77890-220 77890-221 77890-222
7-Chloro-DL-tryptophan ≥95%, Ambeed.Inc
7-Chloro-DL-tryptophan

Precaución: For research use only.

Formula: C₁₁H₁₁ClN₂O₂
MW: 238.67 g/mol
MDL Number: MFCD09264324
CAS Number: 153-97-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR