(2-(Methoxycarbonyl)benzo[b]thiophen-5-yl)boronic acid ≥95%

Supplier: Ambeed
Warning

A2016870-100MG A2016870-250MG
77598-294EA 179.38 USD
77598-294 77626-010
(2-(Methoxycarbonyl)benzo[b]thiophen-5-yl)boronic acid ≥95%
(2-(Methoxycarbonyl)benzo[b]thiophen-5-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₉BO₄S
MW: 236.05 g/mol
MDL Number: MFCD29075230
CAS Number: 1216665-41-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR