(3-Cyano-4-((tetrahydro-2H-pyran-4-yl)oxy)phenyl)boronic acid 95+%

Supplier: Ambeed
Warning

A2014675-100MG A2014675-1G A2014675-250MG A2014675-5G
77877-607EA 70 USD
77877-607 77877-608 77877-609 77877-610
(3-Cyano-4-((tetrahydro-2H-pyran-4-yl)oxy)phenyl)boronic acid 95+%
(3-Cyano-4-((tetrahydro-2H-pyran-4-yl)oxy)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₄BNO₄
MW: 247.05 g/mol
MDL Number: MFCD28147899
CAS Number: 876918-86-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR