2-Bromo-3-chlorobenzonitrile ≥95%

Supplier: Ambeed
Warning

A201125-100MG A201125-250MG A201125-5G
77901-587EA 16 USD
77901-587 77901-103 77900-899
2-Bromo-3-chlorobenzonitrile ≥95%
2-Bromo-3-chlorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrClN
MDL Number: MFCD10699544
CAS Number: 1031929-33-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR