2-Methoxyestradiol ≥98%

Supplier: Ambeed
Danger

A200225-100MG A200225-250MG A200225-50MG
77615-698EA 53.31 USD
77615-698 77615-700 77748-265
2-Methoxyestradiol ≥98%
2-Methoxyestradiol

Precaución: for Research Use Only

Formula: C₁₉H₂₆O₃
MDL Number: MFCD00010489
CAS Number: 362-07-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR