Ethyl benzenesulfonate ≥98%

Supplier: Ambeed
Warning

Synonyms: Benzenesulphonic Acid Ethyl Ester

A200118-25G A200118-500G A200118-100G
77625-990EA 27.81 USD
77625-990 77877-587 77877-586
Ethyl benzenesulfonate ≥98%
Ethyl benzenesulfonate

Precaución: for Research Use Only

Formula: C₈H₁₀O₃S
MW: 186.23 g/mol
MDL Number: MFCD00025033
CAS Number: 515-46-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR