6-Chloro-1-hexyne ≥98%

Supplier: Ambeed
Danger

A199989-1G A199989-25G A199989-5G
77611-928EA 22.13 USD
77611-928 77611-930 77611-932
6-Chloro-1-hexyne ≥98%
6-Chloro-1-hexyne

Precaución: for Research Use Only

Formula: C₆H₉Cl
MDL Number: MFCD00013697
CAS Number: 10297-06-0
UN: 1993
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR